(2-Chloro-6-fluoro-3-methoxyphenyl)acetic acid 98%
Supplier: Apollo Scientific
PC302447-1G
PC302447-5G
PC302447-10G
APOSPC302447-1GEA
144
GBP
APOSPC302447-1G
APOSPC302447-5G
APOSPC302447-10G
(2-Chloro-6-fluoro-3-methoxyphenyl)acetic acid 98%
(2-Chloro-6-fluoro-3-methoxyphenyl)acetic acid
Formula:
C₉H₈ClFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09832268 CAS Number: 1017777-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...