3-Ethoxy-2,6-difluorobenzaldehyde 95%

Supplier: Apollo Scientific

PC302295-5G PC302295-10G PC302295-1G
APOSPC302295-5GEA 83.8 GBP
APOSPC302295-5G APOSPC302295-10G APOSPC302295-1G
3-Ethoxy-2,6-difluorobenzaldehyde 95%
3-Ethoxy-2,6-difluorobenzaldehyde
Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11519366
CAS Number: 1028263-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR