4-Chloro-2-fluoro-3-methoxycinnamic acid 97%

Supplier: Apollo Scientific

Synonyms: 4-Chloro-2-fluoro-3-methoxycinnamic acid

PC302206-1G PC302206-5G
APOSPC302206-1GEA 73.5 GBP
APOSPC302206-1G APOSPC302206-5G
4-Chloro-2-fluoro-3-methoxycinnamic acid 97%
4-Chloro-2-fluoro-3-methoxycinnamic acid
Formula: C₁₀H₈ClFO₃
MW: 230,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687212
CAS Number: 1353001-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR