(4-Chloro-2-(trifluoromethyl)phenyl)acetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC302058-1G
PC302058-5G
PC302058-10G
APOSPC302058-1GEA
83.8
GBP
APOSPC302058-1G
APOSPC302058-5G
APOSPC302058-10G
(4-Chloro-2-(trifluoromethyl)phenyl)acetic acid 98%
(4-Chloro-2-(trifluoromethyl)phenyl)acetic acid
Formula:
C₉H₆ClF₃O₂ MW: 238.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115842 CAS Number: 601513-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...