4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride

Supplier: Apollo Scientific
Danger

PC300003-1G PC300003-250MG PC300003-500MG
APOSPC300003-1GEA 201 GBP
APOSPC300003-1G APOSPC300003-250MG APOSPC300003-500MG
4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Formula: C₁₁H₁₃ClFN
MW: 213.68 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00035278
CAS Number: 1978-61-6
EINECS: 217-836-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR