(2,5-Difluorophenyl)acetic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2,5-Difluorophenylacetic acid

PC2874C-25G PC2874C-5G
APOSPC2874C-25GEA 59.1 GBP
APOSPC2874C-25G APOSPC2874C-5G
(2,5-Difluorophenyl)acetic acid 97%
(2,5-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00010000
CAS Number: 85068-27-5
EINECS: 285-288-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR