5-Fluoro-2-methoxybenzyl bromide 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(Bromomethyl)-4-fluoro-1-methoxybenzene
PC2832-1G
PC2832-25G
PC2832-5G
APOSPC2832-1GEA
14.5
GBP
APOSPC2832-1G
APOSPC2832-25G
APOSPC2832-5G
5-Fluoro-2-methoxybenzyl bromide 97%
5-Fluoro-2-methoxybenzyl bromide
Formula:
C₈H₈BrFO MW: 219,05 g/mol Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671769 CAS Number: 700381-18-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...