2-Chloro-4-fluoro-5-nitrotoluene
Supplier: Apollo Scientific
|
Warning
|
PC27850-5G
PC27850-25G
APOSPC27850-5GEA
19.7
GBP
APOSPC27850-5G
APOSPC27850-25G
2-Chloro-4-fluoro-5-nitrotoluene
2-Chloro-4-fluoro-5-nitrotoluene
Formula:
C₇H₅ClFNO₂ Melting Pt: 40…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD11110549 CAS Number: 112108-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...