2,3-Difluorobenzophenone 97%

Supplier: Apollo Scientific

PC2689-10G PC2689-50G PC2689-5G PC2689-100G
APOSPC2689-10GEA 76.1 GBP
APOSPC2689-10G APOSPC2689-50G APOSPC2689-5G APOSPC2689-100G
2,3-Difluorobenzophenone 97%
2,3-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218,2 g/mol
Boiling Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00061139
CAS Number: 208173-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR