2',5'-Difluoroacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2,5-Difluorophenyl)ethanone, 1-(2,5-Difluorophenyl)ethan-1-one
PC2576-100G
PC2576-25G
PC2576-5G
APOSPC2576-100GEA
85.2
GBP
APOSPC2576-100G
APOSPC2576-25G
APOSPC2576-5G
2',5'-Difluoroacetophenone 98%
2',5'-Difluoroacetophenone
Formula:
C₈H₆F₂O MW: 156,13 g/mol Boiling Pt: 190…191 °C Density: 1,233 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009898 CAS Number: 1979-36-8 EINECS: 217-837-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...