3-Chloro-2,4-difluorobenzaldehyde 97%

Supplier: Apollo Scientific
Warning

PC2571-10G PC2571-5G
APOSPC2571-10GEA 296 GBP
APOSPC2571-10G APOSPC2571-5G
3-Chloro-2,4-difluorobenzaldehyde 97%
3-Chloro-2,4-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116001
CAS Number: 127675-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR