2-(Trifluoromethoxy)terephthalonitrile 98%
Supplier: Apollo Scientific
PC2562-1G
PC2562-5G
PC2562-10G
APOSPC2562-1GEA
93.1
GBP
APOSPC2562-1G
APOSPC2562-5G
APOSPC2562-10G
2-(Trifluoromethoxy)terephthalonitrile 98%
2-(Trifluoromethoxy)terephthalonitrile
Formula:
C₉H₃F₃N₂O MW: 212,131 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00203479 CAS Number: 175278-16-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...