1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

PC2202-5G PC2202-1G
APOSPC2202-5GEA 366 GBP
APOSPC2202-5G APOSPC2202-1G
1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid 97%
1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Formula: C₁₁H₆F₃N₃O₄
MW: 301,18 g/mol
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00173867
CAS Number: 142818-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR