4-(4-Fluorophenyl)piperidine hydrochloride 97%
Supplier: Apollo Scientific
PC201277-10G
PC201277-5G
APOSPC201277-10GEA
242
GBP
APOSPC201277-10G
APOSPC201277-5G
4-(4-Fluorophenyl)piperidine hydrochloride 97%
4-(4-Fluorophenyl)piperidine hydrochloride
Formula:
C₁₁H₁₅ClFN MW: 215,7000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01632528 CAS Number: 6716-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...