5-Cyano-2-fluorobenzaldehyde 95+%

Supplier: Apollo Scientific

Synonyms: 4-Fluoro-3-formylbenzonitrile

PC201142-10G PC201142-25G PC201142-5G
APOSPC201142-10GEA 107 GBP
APOSPC201142-10G APOSPC201142-25G APOSPC201142-5G
5-Cyano-2-fluorobenzaldehyde 95+%
5-Cyano-2-fluorobenzaldehyde
Formula: C₈H₄FNO
MW: 149,1219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974126
CAS Number: 146137-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR