2-Amino-3,5-difluorobenzonitrile 97%

Supplier: Apollo Scientific

PC201107-10G PC201107-1G
APOSPC201107-10GEA 364 GBP
APOSPC201107-10G APOSPC201107-1G
2-Amino-3,5-difluorobenzonitrile 97%
2-Amino-3,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18826239
CAS Number: 126674-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR