5-Bromo-2-(4-fluorophenyl)pyridine

Supplier: Apollo Scientific

PC200356-1G PC200356-500MG
APOSPC200356-1GEA 656 GBP
APOSPC200356-1G APOSPC200356-500MG
5-Bromo-2-(4-fluorophenyl)pyridine
5-Bromo-2-(4-fluorophenyl)pyridine
Formula: C₁₁H₇BrFN
MW: 252,0824 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100950
CAS Number: 463336-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR