Potassium pyrimidine-5-trifluoroborate

Supplier: Apollo Scientific

Synonyms: Potassium trifluoro(pyrimidin-5-yl)borate

PC200342-1G PC200342-500MG
APOSPC200342-1GEA 328 GBP
APOSPC200342-1G APOSPC200342-500MG
Potassium pyrimidine-5-trifluoroborate
Potassium pyrimidine-5-trifluoroborate
Formula: C₄H₃BF₃KN₂
MW: 185,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09992922
CAS Number: 1242733-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR