Ethyl-3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Supplier: Apollo Scientific
Warning

PC200212-1G PC200212-500MG PC200212-5G
APOSPC200212-1GEA 165 GBP
APOSPC200212-1G APOSPC200212-500MG APOSPC200212-5G
Ethyl-3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈ClF₃N₂O₂
Melting Pt: 99…100 °C
Storage Temperature: Ambient
MDL Number: MFCD11841001
CAS Number: 1135283-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR