Potassium-4-(hydroxymethyl)phenyltrifluoroborate

Supplier: Apollo Scientific

Synonyms: Potassium trifluoro(4-(hydroxymethyl)phenyl)borate

PC200164-1G PC200164-5G
APOSPC200164-1GEA 55.1 GBP
APOSPC200164-1G APOSPC200164-5G
Potassium-4-(hydroxymethyl)phenyltrifluoroborate
Potassium 4-(hydroxymethyl)phenyltrifluoroborate
Formula: C₇H₇BF₃KO
MW: 214,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276800
CAS Number: 1015082-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR