4'-(Trifluoromethoxy)biphenyl-3-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
PC200127-1G
PC200127-5G
APOSPC200127-1GEA
145
GBP
APOSPC200127-1G
APOSPC200127-5G
4'-(Trifluoromethoxy)biphenyl-3-carboxylic acid
4'-(Trifluoromethoxy)biphenyl-3-carboxylic acid
Formula:
C₁₄H₉F₃O₃ MW: 282,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04117416 CAS Number: 728919-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...