4-Cyano-4-(4-fluorophenyl)cyclohexanone

Supplier: Apollo Scientific

Synonyms: 1-(4-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile

PC200019-1G PC200019-500MG
APOSPC200019-1GEA 151 GBP
APOSPC200019-1G APOSPC200019-500MG
4-Cyano-4-(4-fluorophenyl)cyclohexanone
4-Cyano-4-(4-fluorophenyl)cyclohexanone
Formula: C₁₃H₁₂FNO
MW: 217,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044815
CAS Number: 56326-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR