2-Fluorophenylhydrazine

Supplier: Apollo Scientific
Warning

PC1759-10G PC1759-1G PC1759-5G
APOSPC1759-10GEA 185 GBP
APOSPC1759-10G APOSPC1759-1G APOSPC1759-5G
2-Fluorophenylhydrazine
2-Fluorophenylhydrazine
Formula: C₆H₇FN₂
Boiling Pt: 76…79 °C/3mmHg
Melting Pt: 47 °C
Storage Temperature: Refrigerator
CAS Number: 2368-80-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR