4'-Chloro-2',5'-difluoroacetophenone 97%
Supplier: Apollo Scientific
|
Warning
|
PC1753C-1G
PC1753C-5G
APOSPC1753C-1GEA
129
GBP
APOSPC1753C-1G
APOSPC1753C-5G
4'-Chloro-2',5'-difluoroacetophenone 97%
4'-Chloro-2',5'-difluoroacetophenone
Formula:
C₈H₅ClF₂O MW: 190,58 g/mol Boiling Pt: 90 °C/5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00068247 CAS Number: 655-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...