Potassium (4-methylphenyl)trifluoroborate 96%

Supplier: Apollo Scientific
Warning

Synonyms: Potassium trifluoro(p-tolyl)borate, Potassium p-tolyltrifluoroborate, (4-Methylphenyl)trifluoroboric acid potassium salt

PC1697-1G PC1697-25G PC1697-5G
APOSPC1697-1GEA 52.5 GBP
APOSPC1697-1G APOSPC1697-25G APOSPC1697-5G
Potassium (4-methylphenyl)trifluoroborate 96%
Potassium (4-methylphenyl)trifluoroborate
Formula: C₇H₇BF₃K
MW: 198,04 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318171
CAS Number: 216434-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR