1-Chloro-3-fluoro-2-nitrobenzene
Supplier: Apollo Scientific
|
Warning
|
PC1499-1G
PC1499-25G
PC1499-5G
APOSPC1499-1GEA
19.7
GBP
APOSPC1499-1G
APOSPC1499-25G
APOSPC1499-5G
1-Chloro-3-fluoro-2-nitrobenzene
1-Chloro-3-fluoro-2-nitrobenzene
Formula:
C₆H₃ClFNO₂ Boiling Pt: 148…152 °C/30mmHg Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD06658264 CAS Number: 64182-61-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...