2-Chloro-4'-fluoroacetophenone 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone
PC1463-100G
PC1463-25G
PC1463-500G
APOSPC1463-100GEA
22.3
GBP
APOSPC1463-100G
APOSPC1463-25G
APOSPC1463-500G
2-Chloro-4'-fluoroacetophenone 99%
2-Chloro-4'-fluoroacetophenone
Formula:
C₈H₆ClFO MW: 172,59 g/mol Boiling Pt: 247 °C Melting Pt: 46…50 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011652 CAS Number: 456-04-2 EINECS: 207-256-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...