2'-Bromo-4'-fluoroacetanilide 96%

Supplier: Apollo Scientific

Synonyms: N-(2-Bromo-4-fluorophenyl)acetamide

PC1394M-100G PC1394M-500G
APOSPC1394M-100GEA 145 GBP
APOSPC1394M-100G APOSPC1394M-500G
2'-Bromo-4'-fluoroacetanilide 96%
2'-Bromo-4'-fluoroacetanilide
Formula: H₃CC(O)NH(C₆H₃BrF)
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00061118
CAS Number: 1009-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR