2-Fluoro-5-methylbenzaldehyde
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Fluoro-5-tolualdehyde, 6-Fluoro-m-tolylaldehyde, 6-Fluoro-m-tolualdehyde
PC1373-1G
PC1373-5G
PC1373-10G
APOSPC1373-1GEA
19.7
GBP
APOSPC1373-1G
APOSPC1373-5G
APOSPC1373-10G
2-Fluoro-5-methylbenzaldehyde
2-Fluoro-5-methylbenzaldehyde
Formula:
C₈H₇FO Boiling Pt: 89 °C/15mmHg Density: 1,129 g/cm³ (25 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093762 CAS Number: 93249-44-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...