N-(3-Trifluoromethylphenyl)anthranilic acid 98%

Supplier: Apollo Scientific

Synonyms: Flufenamic acid, 2-[(3-Trifluoromethylphenyl)amino]benzoic acid, 2-(3-Trifluoromethylanilino)benzoic acid, 2-((3-(Trifluoromethyl)phenyl)amino)benzoic acid

PC1352-1KG PC1352-250G
APOSPC1352-1KGEA 519 GBP
APOSPC1352-1KG APOSPC1352-250G
N-(3-Trifluoromethylphenyl)anthranilic acid 98%
N-(3-Trifluoromethylphenyl)anthranilic acid
Formula: C₁₄H₁₀F₃NO₂
MW: 281,23 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002422
CAS Number: 530-78-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR