Flutamide
Supplier: Apollo Scientific
Synonyms:
N-(4-Nitro-3-(trifluoromethyl)phenyl)isobutyramide, 4'-Nitro-3'-(trifluoromethyl)isobutyranilide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide
PC1192-25G
PC1192-5G
APOSPC1192-25GEA
66.9
GBP
APOSPC1192-25G
APOSPC1192-5G
Flutamide
Flutamide
Formula:
C₁₁H₁₁F₃N₂O₃ MW: 276,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00072009 CAS Number: 13311-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...