2-Bromo-4-(trifluoromethyl)benzoic acid 97+%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid, 2-Bromo-4-(trifluoromethl)benzoic acid

PC11107-1G PC11107-5G
APOSPC11107-1GEA 17.1 GBP
APOSPC11107-1G APOSPC11107-5G
2-Bromo-4-(trifluoromethyl)benzoic acid 97+%
2-Bromo-4-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368837
CAS Number: 328-89-2
EINECS: 206-339-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR