2-Amino-5-(trifluoromethyl)pyridine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 5-(Trifluoromethyl)pyridin-2-amine, 5-(Trifluoromethyl)-2-pyridylamine

PC1100H-5G PC1100H-25G
APOSPC1100H-5GEA 23.7 GBP
APOSPC1100H-5G APOSPC1100H-25G
2-Amino-5-(trifluoromethyl)pyridine 98%
2-Amino-5-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162,11 g/mol
Boiling Pt: 90…92 °C/15mmHg
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00042164
CAS Number: 74784-70-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR