5-Amino-α,α,α-trifluoro-m-toluic acid 97%

Supplier: Apollo Scientific
Warning

PC1100CD-100G PC1100CD-25G PC1100CD-5G
APOSPC1100CD-100GEA 455 GBP
APOSPC1100CD-100G APOSPC1100CD-25G APOSPC1100CD-5G
5-Amino-α,α,α-trifluoro-m-toluic acid 97%
5-Amino-α,α,α-trifluoro-m-toluic acid
Formula: C₈H₆F₃NO₂
MW: 205,14 g/mol
Melting Pt: 141…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00236641
CAS Number: 328-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR