2-Fluoro-5-(trifluoromethoxy)benzyl alcohol

Supplier: Apollo Scientific

PC1093-10G PC1093-5G
APOSPC1093-10GEA 215 GBP
APOSPC1093-10G APOSPC1093-5G
2-Fluoro-5-(trifluoromethoxy)benzyl alcohol
2-Fluoro-5-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04115889
CAS Number: 344411-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR