2-Bromo-5-(trifluoromethoxy)phenylacetic acid 99%
Supplier: Apollo Scientific
|
Warning
|
PC1081-1G
PC1081-5G
APOSPC1081-1GEA
112
GBP
APOSPC1081-1G
APOSPC1081-5G
2-Bromo-5-(trifluoromethoxy)phenylacetic acid 99%
2-Bromo-5-(trifluoromethoxy)phenylacetic acid
Formula:
C₉H₆BrF₃O₃ Storage Temperature: Ambient |
CAS Number:
887266-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...