6-Fluoroanthranilic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-6-fluorobenzoic acid
PC1071M-100G
PC1071M-25G
APOSPC1071M-100GEA
28.8
GBP
APOSPC1071M-100G
APOSPC1071M-25G
6-Fluoroanthranilic acid 97%
6-Fluoroanthranilic acid
Formula:
C₇H₆FNO₂ Melting Pt: 165…172 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00067781 CAS Number: 434-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...