3,5-Difluoropyridine 98%
Supplier: Apollo Scientific
|
Danger
|
PC10528-1G
PC10528-5G
APOSPC10528-1GEA
13.2
GBP
APOSPC10528-1G
APOSPC10528-5G
3,5-Difluoropyridine 98%
3,5-Difluoropyridine
Formula:
C₅H₃F₂N MW: 115,08 g/mol Boiling Pt: 92…93 °C Density: 1,268 g/cm³ (25 °C) Flash Pt: 7,2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03412222 CAS Number: 71902-33-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...