3,5-Difluoropyridine-2-carbonitrile

Supplier: Apollo Scientific
Warning

PC10521-25G PC10521-5G
APOSPC10521-25GEA 89.1 GBP
APOSPC10521-25G APOSPC10521-5G
3,5-Difluoropyridine-2-carbonitrile
3,5-Difluoropyridine-2-carbonitrile
Formula: C₆H₂F₂N₂
Melting Pt: 30…34 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788759
CAS Number: 298709-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR