4-Bromo-3-fluoropyridine

Supplier: Apollo Scientific
Warning

PC10415-1G PC10415-250MG PC10415-5G
APOSPC10415-1GEA 61.7 GBP
APOSPC10415-1G APOSPC10415-250MG APOSPC10415-5G
4-Bromo-3-fluoropyridine
4-Bromo-3-fluoropyridine
Formula: C₅H₃BrFN
MW: 175,9864 g/mol
Density: 1,74 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD04113266
CAS Number: 2546-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR