Methyl-5-fluorosalicylate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 5-fluoro-2-hydroxybenzoate
PC10377-1G
PC10377-25G
PC10377-5G
APOSPC10377-1GEA
44.6
GBP
APOSPC10377-1G
APOSPC10377-25G
APOSPC10377-5G
Methyl-5-fluorosalicylate 98%
Methyl 5-fluorosalicylate
Formula:
C₈H₇FO₃ MW: 170,1400 g/mol Boiling Pt: 50 °C/0,15mmHg Melting Pt: 30…34 °C Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427141 CAS Number: 391-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...