3-Amino-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbonitrile
Supplier: Apollo Scientific
Synonyms:
3-Amino-4-{[3-(trifluoromethyl)phenyl]thio}benzonitrile
PC10347-1G
PC10347-500MG
PC10347-5G
APOSPC10347-1GEA
481
GBP
APOSPC10347-1G
APOSPC10347-500MG
APOSPC10347-5G
3-Amino-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbonitrile
3-Amino-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbonitrile
Formula:
C₁₄H₉F₃N₂S MW: 294,3 g/mol Melting Pt: 149…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD03001303 CAS Number: 400751-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...