4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,2-Dihydro-4-(4-fluorophenyl)-2-oxopyridine-3-carbonitrile
PC10344-1G
PC10344-250MG
APOSPC10344-1GEA
315
GBP
APOSPC10344-1G
APOSPC10344-250MG
4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formula:
C₁₂H₇FN₂O MW: 214,2 g/mol Melting Pt: 284…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD04117821 CAS Number: 886360-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...