5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5,7-Bis(trifluoromethyl)-1,8-naphthyridin-2-amine, 7-Amino-2,4-bis(trifluoromethyl)-1,8-naphthyridine
PC10284-1G
PC10284-250MG
PC10284-5G
APOSPC10284-1GEA
91.8
GBP
APOSPC10284-1G
APOSPC10284-250MG
APOSPC10284-5G
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine 95+%
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine
Formula:
C₁₀H₅F₆N₃ MW: 281,16 g/mol Melting Pt: 195…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00140205 CAS Number: 51420-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...