5-Bromo-1-(3-fluorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Supplier: Apollo Scientific

PC10282-1G PC10282-500MG PC10282-5G
APOSPC10282-1GEA 366 GBP
APOSPC10282-1G APOSPC10282-500MG APOSPC10282-5G
5-Bromo-1-(3-fluorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
5-Bromo-1-(3-fluorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formula: C₁₃H₉BrFNO₃
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD05663804
CAS Number: 886360-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR