1-(3-Fluorobenzyl)-4-piperidinecarbohydrazide

Supplier: Apollo Scientific

PC10274-1G PC10274-500MG PC10274-5G
APOSPC10274-1GEA 366 GBP
APOSPC10274-1G APOSPC10274-500MG APOSPC10274-5G
1-(3-Fluorobenzyl)-4-piperidinecarbohydrazide
1-(3-Fluorobenzyl)-4-piperidinecarbohydrazide
Formula: C₁₃H₁₈FN₃O
MW: 251,31 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD03001285
CAS Number: 453557-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR