3-Amino-3-(4-fluorophenyl)propionic acid
Supplier: Apollo Scientific
PC10261-1G
PC10261-5G
PC10261-10G
APOSPC10261-1GEA
48.6
GBP
APOSPC10261-1G
APOSPC10261-5G
APOSPC10261-10G
3-Amino-3-(4-fluorophenyl)propionic acid
3-Amino-3-(4-fluorophenyl)propionic acid
Formula:
C₉H₁₀FNO₂ MW: 183,18 g/mol Melting Pt: 223…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00181811 CAS Number: 325-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...