5-(Carboxymethyl)-1-[4-(trifluoromethoxy)phenyl]-1H-1,2,3-triazole-4-carboxylic acid

Supplier: Apollo Scientific

PC10214-1G PC10214-500MG
APOSPC10214-1GEA 366 GBP
APOSPC10214-1G APOSPC10214-500MG
5-(Carboxymethyl)-1-[4-(trifluoromethoxy)phenyl]-1H-1,2,3-triazole-4-carboxylic acid
5-(Carboxymethyl)-1-[4-(trifluoromethoxy)phenyl]-1H-1,2,3-triazole-4-carboxylic acid
Formula: C₁₂H₈F₃N₃O₅
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD05021855
CAS Number: 439095-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR