(2,2,2-Trifluoroethyl)oxirane
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1,1,1-Trifluoro-3,4-epoxybutane, 1,2-Epoxy-4,4,4-trifluorobutane
PC10086-1G
PC10086-5G
APOSPC10086-1GEA
59.1
GBP
APOSPC10086-1G
APOSPC10086-5G
(2,2,2-Trifluoroethyl)oxirane
(2,2,2-Trifluoroethyl)oxirane
Formula:
C₄H₅F₃O MW: 126,08 g/mol Boiling Pt: 82 °C Density: 1,2487 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00156048 CAS Number: 407-12-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...