1-Iodo-3-(trifluoromethoxy)benzene 97+%

Supplier: Apollo Scientific
Warning

PC0954-25G PC0954-1G
APOSPC0954-25GEA 78.7 GBP
APOSPC0954-25G APOSPC0954-1G
1-Iodo-3-(trifluoromethoxy)benzene 97+%
1-Iodo-3-(trifluoromethoxy)benzene
Formula: C₇H₄F₃IO
MW: 288,01 g/mol
Boiling Pt: 74…76 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD01090992
CAS Number: 198206-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR